Globe Glass™ Volumetric Flasks, To Contain, Globe Scientific
Supplier: Globe Scientific
8200001 8200002 8200005 8200010 8200025 8200050 8200100 8200200 8200250 8200500 8201000 8202000 8230005 8230010 8230020 8230025 8230050 8230100 8230200 8230250 8250010 8250025 8250050 8250100 8250200 8250250 8250500 8251000 8252000
76523-258BX 324.08 CAD
76523-258 76523-260 76523-262 76523-264 76523-266 76523-268 76523-270 76523-272 76523-274 76523-276 76523-278 76523-280 76523-282 76523-284 76523-286 76523-288 76523-290 76523-292 76523-294 76523-296 76523-298 76523-300 76523-302 76523-304 76523-306 76523-308 76523-310 76523-312 76523-314
Globe Glass™ Volumetric Flasks, To Contain, Globe Scientific
Flasks Volumetric Flasks
Globe Glass™ volumetric flasks are calibrated to meet ASTM tolerance specifications for precise measurement in solution preparation and dilution. A diamond etched graduation line indicates the calibrated capacity of each measuring flask. Wide mouth options are available for easier filling and mixing. Globe Glass™ volumetric flasks are offered in sizes ranging from 1 to 2000 ml, and with Class A or Class B calibration tolerances. Class A volumetric flasks include a batch calibration certificate.
  • Calibrated TC; 'To Contain'
  • Class A calibration tolerance for highest accuracy and the most critical laboratory work
  • Class B calibration tolerance for general laboratory work
  • Supplied with glass pennyhead stoppers
  • Diamond etched graduation marks
  • High resistance to chemicals
  • Durable white enamel printing
Globe Glass™ lab glassware is made exclusively from ASTM E438, Type I, Class A, 3.3 borosilicate glass for excellent resistance to thermal shock and high chemical resistance to most acids, alkalis, and solvents. Globe Glass™ lab glassware meets applicable ASTM standards for volume, shape, dimensions, graduation markings, and calibration. These ASTM standards are clearly marked for easy confirmation that each piece of laboratory glassware will meet the needs of laboratory professionals. Computer controlled, automated production lines ensure that this laboratory glassware has consistent wall thickness, flat bottoms free of striations, and precise forming of rims and spouts.
Permanent white enamel markings are ASTM compliant and easy to read. Dual metric scale graduations are included on most items for easy reference while filling or dispensing. A large, convenient marking area is provided for coding or identification. Globe Glass™ laboratory glassware is manufactured under stringent quality controls. Manufacturing facilities are ISO 9001 certified and in-house calibration is performed in an ISO 17025 certified laboratory.
Certifications: Sizes 1 and 2 ml meet ASTM E237 specification. Sizes 5 to 2000 ml meet ASTM E288 specification.
Packaging: GlassGuard™ Packaging is engineered to meet the hazards and rigors of transportation, protecting Globe Glass laboratory glassware during shipment. Robust packaging designs begin at the pack level and extend to the full case packaging. Heavy-duty, multi-layered corrugated container systems meet rigid quality standards and testing protocols to provide exceptional protection from damage, even in the small parcel shipping environment.