Globe Glass™ Media Bottles, Globe Scientific
Supplier: Globe Scientific
8100050 8100100 8100250 8100500 8101000 8102000 8105000 810010K
76523-242BX 155.71 CAD
76523-242 76523-244 76523-246 76523-248 76523-250 76523-252 76523-254 76523-256
Globe Glass™ Media Bottles, Globe Scientific
Bottles Glass Bottles
Globe Glass™ media bottles are designed with a wide mouth for ease of filling, dispensing, and pipetting. A drip-free pour ring and autoclavable polyethylene (PE) screw cap enable the sterilization of media and reagents. Globe Glass™ media bottles are available in sizes ranging from 50 to 10000 ml.
  • ASTM E438, Type I, Class A, borosilicate 3.3 glass
  • GL 32 polyethylene (PE) screw cap on size 50 ml
  • GL 45 polyethylene (PE) screw caps on sizes 100 to 10000 ml
  • Pouring ring for drip-free liquid transfer
  • High resistance to chemicals
  • Dual scale metric graduations; durable white enamel printing
  • Large white marking area
Globe Glass™ lab glassware is made exclusively from ASTM E438, Type I, Class A, 3.3 borosilicate glass for excellent resistance to thermal shock and high chemical resistance to most acids, alkalis, and solvents. Globe Glass™ lab glassware meets applicable ASTM standards for volume, shape, dimensions, graduation markings, and calibration. These ASTM standards are clearly marked for easy confirmation that each piece of laboratory glassware will meet the needs of laboratory professionals. Computer controlled, automated production lines ensure that this laboratory glassware has consistent wall thickness, flat bottoms free of striations, and precise forming of rims and spouts.
Permanent white enamel markings are ASTM compliant and easy to read. Dual metric scale graduations are included on most items for easy reference while filling or dispensing. A large, convenient marking area is provided for coding or identification. Globe Glass™ laboratory glassware is manufactured under stringent quality controls. Manufacturing facilities are ISO 9001 certified and in-house calibration is performed in an ISO 17025 certified laboratory.
Packaging: GlassGuard™ Packaging is engineered to meet the hazards and rigors of transportation, protecting Globe Glass laboratory glassware during shipment. Robust packaging designs begin at the pack level and extend to the full case packaging. Heavy-duty, multi-layered corrugated container systems meet rigid quality standards and testing protocols to provide exceptional protection from damage, even in the small parcel shipping environment.