Globe Glass™ Low Form Griffin Style Beakers, Globe Scientific
Supplier: Globe Scientific
8010010 8010020 8010030 8010050 8010100 8010150 8010250 8010400 8010600 8011000 8012000 8014000
76523-186BX 112.51 CAD
76523-186 76523-188 76523-190 76523-192 76523-194 76523-196 76523-198 76523-200 76523-202 76523-204 76523-206 76523-208
Globe Glass™ Low Form Griffin Style Beakers, Globe Scientific
Beakers
Globe Glass™ beakers are made with uniform wall thickness, flat bottoms, and low expansion 3.3 Borosilicate beaker glass, making them a strong and durable choice with excellent resistance to thermal shock. The lab beakers have beaded rims and formed spouts to facilitate accurate and steady pouring of liquids. Globe Glass™ low form Griffin style graduated beakers are available in sizes from 10 to 4000 ml.
  • Meet ASTM E960 Type I Specifications
  • Formed pour spout for easy pouring
  • High resistance to chemicals
  • Uniform wall thickness
  • Dual metric scale graduations
  • Durable white enamel printing
  • Large white marking area
Globe Glass™ lab glassware is made exclusively from ASTM E438, Type I, Class A, 3.3 borosilicate glass for excellent resistance to thermal shock and high chemical resistance to most acids, alkalis, and solvents. Globe Glass™ lab glassware meets applicable ASTM standards for volume, shape, dimensions, graduation markings, and calibration. These ASTM standards are clearly marked for easy confirmation that each piece of Globe Glass™ laboratory glassware will meet the needs of laboratory professionals. Computer controlled, automated production lines ensure that Globe Glass laboratory glassware has consistent wall thickness, flat bottoms free of striations, and precise forming of rims and spouts. Permanent white enamel markings are ASTM compliant and easy to read. Dual metric scale graduations are included on most items for easy reference while filling or dispensing. Most Globe Glass™ laboratory glassware item includes a large, convenient marking area for coding or identification.
Globe Glass™ laboratory glassware is manufactured under stringent quality controls. Manufacturing facilities are ISO 9001 certified and in-house calibration is performed in an ISO 17025 certified laboratory.
Packaging: GlassGuard™ packaging is engineered to meet the hazards and rigors of transportation, GlassGuard™ packaging protects Globe Glass laboratory glassware during shipment. Robust packaging designs begin at the pack level and extend to the full case packaging. Heavy-duty, multi-layered corrugated container systems meet rigid quality standards and testing protocols to provide exceptional protection from damage, even in the small parcel shipping environment.