AseptiQuik® Sterile Connector Cuffs
Supplier: Colder Products
AQGCUFF AQLCUFF AQSCUFF
76523-384EA 102.08 CAD
76523-384 76523-386 76523-388
AseptiQuik® Sterile Connector Cuffs
Fittings and Connectors
AseptiQuik® Connector Cuffs provide additional support for more robust applications, so users can continue to transfer media easily with these quick and easy to use AseptiQuik® sterile connectors.
  • Genderless design eases single-use system specifications with one-part number for both halves
  • Flip-Click-Pull: intuitive three-step connection process reduces risk of operator error
  • Audible confirmation of connection with no additional hardware required
  • BPA-free
Use the AseptiQuik® cuffs to support use cases which have excessive sideload, pump, tension, vibration, or reduction in flow.
AseptiQuik® PPSU enables genderless connection for a greater range of chemical applications, offering versatile connections across downstream processes with a pH range from 2 to 12.