Globe Glass™ Erlenmeyer Flasks, Globe Scientific
Supplier: Globe Scientific
8400025 8400050 8400125 8400250 8400300 8400500 8401000 8402000 8410125 8410250 8410500 8411000 8420250 8420500 8421000 8422000
76523-210BX 131.51 CAD
76523-210 76523-212 76523-214 76523-216 76523-218 76523-220 76523-222 76523-224 76523-226 76523-228 76523-230 76523-232 76523-234 76523-236 76523-238 76523-240
Globe Glass™ Erlenmeyer Flasks, Globe Scientific
Flasks Erlenmeyer Flasks
Globe Glass™ Erlenmeyer flasks meet ASTM E1404 specifications for narrow mouth, wide mouth and screw cap options and are available in a wide range of sizes.
  • High resistance to chemicals
  • Uniform wall thickness
  • Durable white enamel printing
  • Large white marking area
  • Narrow and wide mouth options have dual metric scale graduations
  • Screw cap options have single metric scale graduations
Wide mouth conical flasks allow easy access to contents, while screw cap conical flasks include a GL 45 PE cap with a PTFE liner for a leak resistant closure. Globe GlassMore Product Information