Membrane Disc Filters
Supplier: Sartorius
1260225ALK 1260237ALK 1260247ALK 1260247ALN 1260250ALK 1260250ALN 1260280ALK
CA76518-018PK 327.45 CAD
CA76518-018 CA76518-020 CA76518-022 CA76518-024 CA76518-026 CA76518-028 CA76518-030
Membrane Disc Filters
Filters Membrane Filters
Gelatine membrane filters in conjunction with the MD8 air samplers (gelatine filter method) are used for the collection of airborne microbes and viruses. These filters are completely water-soluble, therefore, microbes in one sample can be cultivated in different nutrient media or low and high bacteria counts can be measured.
  • Pre-sterilized and ready to use
  • Maintains viability of the collected microorganisms
  • Solubility of the filter is a prerequisite for virus sampling
  • Air flow rate: 2.7 L/min/cm² (0.05 bar)
'Absolute' retention rate of 99,9995% for More Product Information