BACTair™ Culture Media, in Petri Dishes, Sartorius
Supplier: SARTORIUS CORPORATION EDI CA
14320110ACD
14321110ACD
CA76518-036PK
122.63
CAD
CA76518-036
CA76518-038
BACTair™ Culture Media, in Petri Dishes, Sartorius
Media for Microbiology
Prepared Media for Microbiology
BACTair™ culture media plates are prefilled agar media plates used for microbiological air monitoring. Airborne microorganisms are directly captured on the culture media plates, where the plates function directly as collection heads. They are supplied prefilled with different media. These plates filled with culture medium should be stored at 15 to 25 °C.
- Sterile and gamma irradiated
- Individually packaged in PE bag
- Integrated disposable sieve
- Size: 90 mm