Lba4404 Chemically Competent Agrobacterium, Intact Genomics
Supplier: Intact Genomics
1085-06 1085-18
104013-292EA 300 CAD
104013-292 104013-294
Lba4404 Chemically Competent Agrobacterium, Intact Genomics
Competent Cells
Intact Genomics LBA4404 Chemically Competent Agrobacterium cells are useful for transgenic operations of tomatoes, tobacco and other plants.
  • High transformation efficiencies
  • Especially useful for cDNA gDNA library construction
Intact Genomics LBA4404 chemically competent Agrobacterium cells are optimized for the highest transformation efficiencies which is ideal for applications requiring high transformation efficiencies, such as with cDNA or gDNA library construction. The LBA4404 strain is useful for transgenic operations of tomatoes, tobacco and other plants. LBA4404 contains a rifampicin resistance gene (rif). LBA4404 strain also contains a octoprine-type Ti plasmid pAL4404 without self-transport function, which contains the vir gene.
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on Friday will be shipped on the following Monday.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.