CrossLab Quick Connect and Quick Turn Fittings for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5067-5957 5067-5958 5067-6166 5500-1172 5500-1173 5500-1188 5500-1189 5500-1191 5500-1193 5067-5959 5067-5960 5067-5961 5067-6167 5067-6602 5500-1170 5500-1171 5500-1179 5500-1180 5500-1181 5500-1182 5500-1183 5500-1190 5500-1192 5500-1194 5500-1195 5500-1196 5500-1197 5500-1198 5500-1205 5500-1230 5500-1232 5500-1233 5500-1235 5500-1237 5500-1247 5067-6163 5067-6168 5067-6169 5500-1174 5500-1178 5500-1206 5500-1231 5500-1234 5500-1236 5500-1248 5500-1276 5500-1288 5500-1289 5500-1290 5500-1596 5500-1597 5500-1599 5067-6164 5067-6165 5500-1175 5500-1176 5500-1177 5500-1277 5500-1291 5500-1416 5500-1474 5500-1578 5500-1581 5500-1582 5500-1583 5500-1584 5500-1585 5500-1586 5500-1587 5500-1590 5500-1591 5500-1592 5500-1593 5500-1594 5500-1595 5500-1598 5500-1600 5500-1601 5500-1602 5500-1603 5500-1604 5500-1605
CA5067-5957EA 588.31 CAD
CA5067-5957 CA5067-5958 CA5067-6166 CA5500-1172 CA5500-1173 CA5500-1188 CA5500-1189 CA5500-1191 CA5500-1193 CA5067-5959 CA5067-5960 CA5067-5961 CA5067-6167 CA5067-6602 CA5500-1170 CA5500-1171 CA5500-1179 CA5500-1180 CA5500-1181 CA5500-1182 CA5500-1183 CA5500-1190 CA5500-1192 CA5500-1194 CA5500-1195 CA5500-1196 CA5500-1197 CA5500-1198 CA5500-1205 CA5500-1230 CA5500-1232 CA5500-1233 CA5500-1235 CA5500-1237 CA5500-1247 CAAG5067-6163 CAAG5067-6168 CAAG5067-6169 CAAG5500-1174 CAAG5500-1178 CAAG5500-1206 CAAG5500-1231 CAAG5500-1234 CAAG5500-1236 CAAG5500-1248 CAAG5500-1276 CAAG5500-1288 CAAG5500-1289 CAAG5500-1290 CAAG5500-1596 CAAG5500-1597 CAAG5500-1599 CAAG5067-6164 CAAG5067-6165 CAAG5500-1175 CAAG5500-1176 CAAG5500-1177 CAAG5500-1277 CAAG5500-1291 CAAG5500-1416 CAAG5500-1474 CAAG5500-1578 CAAG5500-1581 CAAG5500-1582 CAAG5500-1583 CAAG5500-1584 CAAG5500-1585 CAAG5500-1586 CAAG5500-1587 CAAG5500-1590 CAAG5500-1591 CAAG5500-1592 CAAG5500-1593 CAAG5500-1594 CAAG5500-1595 CAAG5500-1598 CAAG5500-1600 CAAG5500-1601 CAAG5500-1602 CAAG5500-1603 CAAG5500-1604 CAAG5500-1605
CrossLab Quick Connect and Quick Turn Fittings for HPLC, Agilent Technologies
Tubing Chromatography Tubing
Finger-tight, reusable fittings for zero-dead-volume connections for your UHPLC.
  • Truly finger-tight connections mean no special training is needed, so differences in user style will not affect your chromatography
  • Spring-loaded design enables zero-dead-volume connections with all types of UHPLC columns as easily as pressing a lever
  • Reliable and reusable InfinityLab Quick Connect fittings are long lasting, reseal tightly, and are stable up to 1300 bar (18850 psi) even after 200+ reconnections
  • Fast, easy connections save time and trouble
  • Winner of the TASIA Award
  • Reduce the need for repetitive wrench work in tight spaces enhancing lab ergonomics
Agilent InfinityLab fittings with innovative spring-loaded design give you the complete confidence of a zero-dead-colume connection, helping you avoid the broad or tailing peaks, loss of resolution, and overall poor chromatographic performance associated with poorly connected fittings. InfinityLab Quick Connect fittings are truly finger tight up to 1300 bar, for column connections as easy as pressing a lever. For harder-to-reach areas on your LC instrument, the InfinityLab Quick Turn fitting produces either a finger-tight connection (up to 600 bar), or a 1300 bar connection with a mounting tool or a wrench.