Peristaltic Pump Tubing for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
3710034400 3710034600 3710049000 3710027200 3710034500 3710034800 3710035000 3710035200 3710046900 3710068100 3710068300 3710068800 5043-0063 9910075900 3710027300 3710043900 3710047000 3710068000 3710068200 3710041700 3710041800 3710041900 3710043500 3710043700 3710043800 3710044000 3710044100 3710044200 3710044300 3710044400 3710053600 3710057500 3710068400 3710068500 3710068600 3710069000 3710069100 3710069300 8003-0459
CA3710034400PK 82.08 CAD
CA3710034400 CA3710034600 CA3710049000 CA3710027200 CA3710034500 CA3710034800 CA3710035000 CA3710035200 CA3710046900 CA3710068100 CA3710068300 CA3710068800 CA5043-0063 CA9910075900 CAAG3710027300 CAAG3710043900 CAAG3710047000 CAAG3710068000 CAAG3710068200 CAAG3710041700 CAAG3710041800 CAAG3710041900 CAAG3710043500 CAAG3710043700 CAAG3710043800 CAAG3710044000 CAAG3710044100 CAAG3710044200 CAAG3710044300 CAAG3710044400 CAAG3710053600 CAAG3710053700 CAAG3710057500 CAAG3710068400 CAAG3710068500 CAAG3710068600 CAAG3710068700 CAAG3710069000 CAAG3710069100 CAAG3710069300 CAAG8003-0459
Peristaltic Pump Tubing for ICP-OES, Agilent Technologies
Tubing Peristaltic Pump Tubing
PVC, Solvaflex, Marprene and FPM peristaltic pump tubing for ICP-OES.
  • Standard PVC suitable for all aqueous with medium concentrated acids or bases
  • PVC Solvaflex for common organic solvents such as kerosene, xylene, white spirit, alcohols, medium-highly concentrated acids and bases
  • Marprene tubing for ketone-based solvents such as MIBK and DIBK;FPM Fluoroelastomer Polymer tubing is suitable for gasoline, naphtha, toluene, xylene (aromatic hydrocarbons) and highly concentrated acids and bases
Agilent top-quality ICP-OES Peristaltic Pump Tubing allows you to find the appropriate product to handle any matrix. Agilent has tubing in the diameters and material required for your application: PVC, Solvaflex, FPM, or Marprene.
;Standard PVC works for all aqueous with medium concentrated acids or bases, PVC Solvaflex works for common organic solvents such as kerosene, xylene and white spirit. Marprene tubing is for ketone-based solvents such as MIBK and DIBK. FPM Fluoroelastomer Polymer tubing is for gasoline, naphtha, toluene, xylene (aromatic hydrocarbons) and highly concentrated acids and bases.