CrossLab Capillary and Tubing Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5065-9937 5065-9939 5061-3304 5067-6614 5067-6615 5067-6707 5004-0003 5013-0002 5061-3315 5065-9938 5067-4234 5067-4249 5067-4250 5067-4251 5067-4252 5067-4270 5067-4294 5067-4300 5067-4633 5067-6181 5067-6187 5067-6596 5067-6597 5067-6598 5067-6697 5067-6708 5067-6800 5067-6802 G1316-68716 G5611-68711 5067-6801 5067-6804
CA5065-9937EA 1729.97 CAD
CA5065-9937 CA5065-9939 CA5061-3304 CAAG5067-6614 CAAG5067-6615 76537-448 CAAG5004-0003 CAAG5013-0002 CAAG5061-3315 CAAG5065-9938 CAAG5067-4234 CAAG5067-4249 CAAG5067-4250 CAAG5067-4251 CAAG5067-4252 CAAG5067-4270 CAAG5067-4294 CAAG5067-4300 CAAG5067-4633 CAAG5067-6181 CAAG5067-6187 CAAG5067-6596 CAAG5067-6597 CAAG5067-6598 CAAG5067-6697 CAAG5067-6708 CAAG5067-6800 CAAG5067-6802 CAAGG131668716 CAAGG561168711 AG5067-6801 AG5067-6804
CrossLab Capillary and Tubing Kits for HPLC, Agilent Technologies
Tubing Chromatography Tubing
A complete set of system capillaries and fittings to efficiently ramp up usage of LC instruments.
  • Carefully selected capillary dimensions (inner diameter and length) to minimize extra-column volume and optimize separation efficiency
  • Peace of mind of having everything needed to make necessary connections on your system
  • InfinityLab Quick Connect and Quick Turn fittings in the InfinityLab capillary kits ensure ease of use and zero dead-volume column connection
Agilent InfinityLab capillary kits are complete sets of system capillaries, includingMore Product Information