CrossLab Ultimate Union for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
2307230001 2307232901 G3182-61580 G3182-60581 2302533140 325-045-HSP 325-062-HSP 325-185-HSP G3182-61581 998-0000053-EHS
CA2307230001EA 352.04 CAD
CA2307230001 CA2307232901 CAG3182-61580 CAG3182-60581 CAAG2302533140 CAAG325-045HSP CAAG325-062HSP CAAG325-185HSP CAAGG318261581 CAG9980000053
CrossLab Ultimate Union for GC, Agilent Technologies
Fittings and Connectors
Ultimate Union kits, part of Agilent's capillary flow technology family, provide extremely low dead volume column connections.
  • Ensure a reliable, robust, and high-quality connection for leak-free results, time after time
  • Eliminate the time-consuming curing step required with press-fit connectors and polyimide glue
  • Strong, reliable, and reusable mechanical seal based on Agilent's exclusive MicroFluidics technology
  • No need to retighten after temperature cycles as with other metal union designs
  • Reuse your column union with confidence in having a dependable
More Product Information