CrossLab Graphite Ferrules for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0100-1324 0100-1325 5062-3516 500-2118 5080-8773 500-2114 5190-4054 CR213164 CR213165 CR2111XX
CA0100-1324PK 77.48 CAD
CA0100-1324 CA0100-1325 CA5062-3516 CA76508-032 CA76508-082 CA500-2114 CA5190-4054 CACR213164 CACR213165 AGCR2111XX
CrossLab Graphite Ferrules for GC
Fittings and Connectors
Graphite ferrules can withstand temperatures as high as 450 °C. Their inherent softness makes them ideal for metal columns and allows for additional installation tolerances.
  • General purpose for capillary columns
  • Suitable for FID, NPD, and ECD
  • Recommended for high temperature and cool on-column applications
  • Material is free of sulfur and other contaminants
  • Can be reused or removed easily
  • Proper installation requires a finger-tight turn on the nut, then an additional 1/4 turn with a wrench
  • Not for use with MS or oxygen sensitive detectors