CrossLab Fittings for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0100-1631 5022-6536 5042-6500 5062-8518 5062-8541 5065-4422 5065-4426 5067-1557 5067-1558 5062-2418 0100-1516 01080-83202 5063-6591 5065-4454 5180-4108 5180-4114 0100-1259 5067-4733 5067-4738 5042-8957 5043-0277 5067-6141 UCP316 5067-1540 5067-1547 5067-1585 5067-4739 RE6000282 RE6000282MC UCF130 UCP628 5065-9967 5067-5695 5005-0067 5005-0068 5023-0232 5043-0259 5063-6593 5065-4410 5065-4423 5065-9948 5065-9950 8710-1217 G1314-65062 RF0030T RF0031T RF0032T RF0098T RF0100T
CA0100-1631PK 121 CAD
CA0100-1631 CA5022-6536 CA5042-6500 CA5062-8518 CA5062-8541 CA5065-4422 CA5065-4426 CA5067-1557 CA5067-1558 CA76508-050 CA76514-494 CA76514-512 CA76514-670 CA76514-674 CA76514-718 CA76514-720 CA0100-1259 CA5067-4733 CA5067-4738 CA5042-8957 CA5043-0277 CA5067-6141 CAUCP316 CAAG5067-1540 CAAG5067-1547 CAAG5067-1585 CAAG5067-4739 CAAGRE6000282 CAAGRE600282MC CAAGUCF130 CAAGUCP628 CA5065-9967 CA5067-5695 CAAG5005-0067 CAAG5005-0068 CAAG5023-0232 CAAG5043-0259 CAAG5063-6593 CAAG5065-4410 CAAG5065-4423 CAAG5065-9948 CAAG5065-9950 CAAG8710-1217 CAAGG131465062 CAAGRF0030T CAAGRF0031T CAAGRF0032T CAAGRF0098T CAAGRF0100T
CrossLab Fittings for HPLC, Agilent Technologies
Fittings and Connectors
Fittings made of different materials to ensure reliable sealing and optimal performance in your LC.
  • Innovative InfinityLab Quick Connect and Quick Turn fittings providing finger-tight, reusable, and zero-dead-volume connections
  • A variety of fitting materials are available, including stainless steel, PEEK, and polyketone, for different pressure ranges and positions
  • Different dimensions covering Swagelok-type or metric M4/M3-type connection ports
  • An array of nuts, ferrules, various color identifiers, and other supplies are also available for all fitting types
Agilent has created a large variety of fittings made of different materials to ensure the ongoing security and performance of your LC equipment. Utilizing the correct fitting style and format, as specified by the manufacturer, is important for ensuring accuracy and robust performance in every aspect of your apparatus.;InfinityLab Quick Connect fittings offer truly finger-tight connections up to 1300 bar, for column connections as easy as pressing a lever. For harder-to-reach areas of the LC instrument, try InfinityLab Quick Turn fittings. You can find the right capillaries quickly and easily using our online selection tool.