CrossLab Column Nuts for Third Party GC Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8004-0311 8003-0311 8001-0015 8001-0016 8001-0020 8001-0311 8001-0312 8002-0010 8002-0013 8002-0311 8002-0312 8003-0010 8004-0009 8004-0312
CA8004-0311PK 56.65 CAD
CA8004-0311 CAAG8003-0311 CAAG8001-0015 CAAG8001-0016 CAAG8001-0020 CAAG8001-0311 CAAG8001-0312 CAAG8002-0010 CAAG8002-0013 CAAG8002-0311 CAAG8002-0312 CAAG8003-0010 CAAG8004-0009 CAAG8004-0312
CrossLab Column Nuts for Third Party GC Instruments, Agilent Technologies
Fittings and Connectors
Column nuts for PerkinElmer, Thermo Scientific, Shimadzu, and Varian/Bruker GC systems.
  • Agilent column nuts for various third-party GC systems seamlessly fit with your instrument
One of the most basic tasks in GC, installing the column, can be challenging. The GC oven is a dark, small space, and column installation uses some tiny parts that need to be assembled with precision. Installation of the column into both the inlet and detector must be done properly to get accurate and reproducible results. Agilent offers a selection of capillary column nutsMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable