VWR® 4 ml Screw-Thread Vials, Kits, and Caps
Supplier: VWR
VWR®
66010-562A 66009-880A 82028-438A 66009-876A 82028-440A 46610-739A
82028-440ACS 614.51 CAD
82028-440A 66009-876A 82028-438A 46610-739A 66010-562A 66009-880A
VWR® 4 ml Screw-Thread Vials, Kits, and Caps
Vials
4 ml screw thread vials are designed for use with Waters® WISP® (48-position) and Shimadzu™ autosamplers.
  • Vials are precision manufactured from high-purity borosilicate glass
  • 13-425 Caps and vials matched for best closure integrity
  • Available with or without writing patch
Vials and caps available as stand alone items or in a convenience kit configuration.