VWR® Screw-Thread Vials 8-425, Caps and Inserts with Standard Opening
Supplier: VWR International
46610-736A 66009-874A 66009-878A 66020-950A 82028-420A 82028-422A
46610-736ACS 500.96 CAD
46610-736A 66009-874A 66009-878A 66020-950A 82028-420A 82028-422A
VWR® Screw-Thread Vials 8-425, Caps and Inserts with Standard Opening
Vials
8-425 autosampler screw thread vials feature a standard opening design compatible with 5 mm inserts.
  • Vials are precision manufactured from high-purity borosilicate glass
  • Caps and vials matched for best closure integrity
  • Available with or without writing patch
  • Spring promotes alignment and ensures Insert seals against septa to help guard against evaporation loss
  • Available pre-assembled, certified, clear glass
MS certified vial kits are tested and certified to meet or exceed up to 14 critical performance parameters. Loose caps and inserts come packaged in poly bags. Polypropylene screw closures available with red PTFE/white silicone septa.
Ordering information: Purchase vial and caps in convenience kits or separately.