CrossLab Ultra-Microcells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5062-2496 5063-6577 6610013700
CA5062-2496EA 903.4 CAD
CA5062-2496 CA5063-6577 CAAG5062-2497 CAAG6610013700
CrossLab Ultra-Microcells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
Ultra-micro rectangular cells with 0.1 to 10 mm path length and 15/20 mm height for to suit 8453/8454/Cary UV.
  • Ultra-micro cells are designed for use in the µl range
  • Fits any standard cell holder; Outstanding measurement reproducibility
  • Ultra-micro cells feature non transmitting, self masking black walls which improve sesitivity by eliminating stray light from measurements
  • Available with 15 or 20 mm z height for compatibility with both 8453/8454 and Cary UV systems
  • Exceptional accuracy in terms of optical path length and parallelism
  • Unique surface flatness of the optical windows
  • High temperature stability; high chemical resistance
  • Guaranteed transmission of >80% from 200 - 3500 nm (depending on the cell window material)
Ultra-micro rectangular cuvettes (cells) are specifically designed for use in the µl range (down to 50 µl) and will fit any standard cell holder. The cells are constructed so that filling and emptying can be easily accomplished with commonly available pipette tips.