CrossLab Rectangular, Triangular and Flow Cells for Fluorescence Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
6610000900 6610001200 6610021200 6610021400 6610021500 6610021600 6610023500 6610023700 6610024200
CAAG6610000900PK 890.9 CAD
CAAG6610000900 CAAG6610001200 CAAG6610021200 CAAG6610021400 CAAG6610021500 CAAG6610021600 CAAG6610023500 CAAG6610023700 CAAG6610024200
CrossLab Rectangular, Triangular and Flow Cells for Fluorescence Systems, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
Fluorescence cells for Cary Eclipse including rectangular, triangular, or flow through cells.
  • Rectangular cells have 4 polished faces for high precision fluorescence measurements
  • Mirrored rectangular cells have mirroring on two adjacent sides for increased sensitivity with cuvette-based fluorescence measurements
  • Fits any standard cell holder and has unique surface flatness of the optical windows, high temperature stability, and high chemical resistance
  • 20mm z height for compatibility with Cary Eclipse fluorescence systems
  • Exceptional accuracy in terms of
More Product Information