CrossLab Flow Cells for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
79883-68703 G1314-65050 5500-1342 5500-1343 5500-1344 5500-1345 G1314-60024 G1314-60025 G1314-60189 G1315-67305 G1315-80001 G1315-87101 G7115-60001 G7115-60002
CA79883-68703EA 348.71 CAD
CA79883-68703 CAG1314-65050 CAAG5500-1342 CAAG5500-1343 CAAG5500-1344 CAAG5500-1345 CAAGG131460024 CAAGG131460025 CAAGG131460189 CAAGG131567305 CAAGG131580001 CAAGG131580002 CAAGG131587101 CAAGG711560001 CAAGG711560002
CrossLab Flow Cells for HPLC, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
High-quality flow cells for Agilent HPLC detectors to allow the best performance in all applications.
  • Designed for highest chromatographic performance
  • Flow cells for nano flow applications
Agilent provides a comprehensive range of high-quality flow cells for Agilent HPLC detectors to allow the best performance in all applications. Available solutions address the requirements of protein research applications or assays to assess the integrity of bio-therapeutic compounds. Bio-inert flow cells are available for Agilent multiple wavelength and diode array UV-absorption detectors, as well as for special detection techniques such as fluorescence and dual-angle light scattering.