Disposable Polystyrene Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
6610018800 6610018700
CA6610018800PK 183.53 CAD
CA6610018800 CAAG6610018700
Disposable Polystyrene Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
Disposable plastic standard or micro cells are economical alternatives to quartz or glass cells.
  • Eliminate the risk of cross-contamination with traditional quartz reusable cuvettes
  • Improve productivity and reduce cost - eliminate the need for cleaning cuvettes for re-use
  • Universal - compatible with virtually all UV/VIS systems without adapters
  • Available in 2 sizes, both with 10 mm path length, 3.5 ml (standard) and 1.5 ml (semi-micro)
  • Usable over wavelength range from 340 - 900 nm
  • Packed in 5 styrofoam trays, each tray holding 100 cells
  • Not recommended for
More Product Information