Top-DNA and TOP-RNA, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
7572915C 75719025 75719050
CAAG7572915CPK 818.66 CAD
CAAG7572915C CAAG75719025 CAAG75719050
Top-DNA and TOP-RNA, Agilent Technologies
Tubes Test Tubes
TOP-DNA and TOP-RNA enable good recovery of high-purity synthetic DNA and RNA oligonucleotides.
  • Enhanced laboratory productivity, a complete solution for DNA and RNA oligonucleotide purification
  • Pre-HPLC 'sample preparation' ability maximizes utility
  • Freeing up operator time, TOP-DNA and TOP-RNA are high-throughput solutions that are automation friendly
  • Reduces operating costs through lower reagent use
  • Gravity or vacuum flow ensures flexibility
Agilent TOP-DNA and TOP-RNA remove interfering salts, incomplete synthesis products, and other impuritiesMore Product Information