Standard Bare Fused Silica Capillaries, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1600-60211 G1600-61211 G1600-62211 G1600-62311 G1600-63211 G1600-60311 G1600-60411 G1600-61311 G1600-62411 G1600-63311 G1600-64211 G1600-64311 G1600-67311
CAG1600-60211PK 206.18 CAD
CAG1600-60211 CAG1600-61211 CAG1600-62211 CAG1600-62311 CAG1600-63211 CAAGG160060311 CAAGG160060411 CAAGG160061311 CAAGG160062411 CAAGG160063311 CAAGG160064211 CAAGG160064311 CAAGG160067311
Standard Bare Fused Silica Capillaries, Agilent Technologies
Electrophoresis Systems Capillary Electrophoresis Systems
Pre-aligned fused-silica capillaries from Agilent Technologies are designed and optimised for ease of use and reliability. All capillary ends are cut to a smooth, mirror-like finish. In addition, the polyimide outer coating is removed from the ends. These processes ensure minimal sample adsorption and help maintain sharp peak shapes. All capillaries have a pre-made detection 'window' and a built-in alignment stopper that allows rapid and precise insertion in the alignment interface.