Single Element Hollow Cathode Lamps, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5610101400 5610102100 5610102700 5610102900 5610124000 5610124800 5610100100 5610100200 5610100300 5610100800 5610101000 5610101200 5610102190 5610103200 5610103400 5610103700 5610104200 5610105000 5610105200 5610105300 5610106800 5610122200 5610123300 5610124600 5610127100 5610127200 5610127300 5610128800
CA5610101400EA 954.04 CAD
CA5610101400 CA5610102100 CA5610102700 CA5610102900 CA5610124000 CA5610124800 CAAG5610100100 CAAG5610100200 CAAG5610100300 CAAG5610100800 CAAG5610101000 CAAG5610101200 CAAG5610102190 CAAG5610103200 CAAG5610103400 CAAG5610103700 CAAG5610104200 CAAG5610105000 CAAG5610105200 CAAG5610105300 CAAG5610106800 CAAG5610122200 CAAG5610123300 CAAG5610124600 CAAG5610127100 CAAG5610127200 CAAG5610127300 CAAG5610128800
Single Element Hollow Cathode Lamps, Agilent Technologies
Lights Hollow Cathode Lamps
Coded and uncoded single element lamps for optimum signal-to-noise ratio and stable performance.
  • Compatible with all Agilent and most AA systems from other manufacturers (except PerkinElmer)
  • Available as either coded or uncoded lamps
  • Easy to use – use our element coded lamps for automatic lamp recognition, preventing errors
  • High performance – manufactured from the purest cathode materials
  • Stable operation – extended processing ensures minimal warm-up and good lamp stability
  • Manufactured in an ISO 9001 certified environment
  • Proven processing steps ensure
More Product Information