Polyvinyl Alcohol (PVA) Coated Capillaries, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1600-61219 G1600-67219 G160U-61219 G1600-60419 G1600-61419 G1600-67319 G1600-68319 G160U-61419
CAG1600-61219EA 535.93 CAD
CAG1600-61219 CAG1600-67219 CAG160U-61219 CAAGG160060419 CAAGG160061419 CAAGG160067319 CAAGG160068319 CAAGG160U61419
Polyvinyl Alcohol (PVA) Coated Capillaries, Agilent Technologies
Electrophoresis Systems Capillary Electrophoresis Systems
PVA coated capillaries contain a permanently adsorbed layer of polyvinyl alcohol.
  • Minimizes hydrophobic and electrostatic solute/wall interactions
  • Eliminates electroosmotic flow (EOF)
  • Capillary types available in greater lengths for use in non-Agilent systems
  • PVA is also available for use with the High-Sensitivity Detection Cell
  • PVA-coated capillaries are also offered for CE/MS applications
Agilent provides a broad range of stable polyvinyl alcohol (PVA)-coated capillaries over a wide pH range, from pH 2.5 to 9.5, allowing the use of a wide range ofMore Product Information