Multi-Element Hollow Cathode Lamps, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5610107600 5610107000 5610108700 5610108800 5610109500
CA5610107600EA 997.12 CAD
CA5610107600 CAAG5610107000 CAAG5610108700 CAAG5610108800 CAAG5610109500
Multi-Element Hollow Cathode Lamps, Agilent Technologies
Lights Hollow Cathode Lamps
Coded and uncoded multi-element lamps allow the same lamp to be used for multiple elements.
  • Compatible with all Agilent and most AA systems from other manufacturers (except PerkinElmer)
  • Easy to use - use our element coded lamps for automatic lamp recognition, preventing errors
  • Save time by eliminating lamp warm-up, when switching elements
  • Same excellent performance as provided by Agilent single element lamps
  • Primary lines checked to avoid and minimize spectral interferences
  • High performance – manufactured from the purest cathode materials
  • Stable operation –
More Product Information