Liner O-Rings for Third Party GC Instruments, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8003-0205 8004-0201 8001-0201 8003-0203 8004-0204 8010-0401 8001-0202 8001-0203 8002-0201 8002-0203 8002-0204 8003-0202 8003-0204 8004-0202 8004-0203
CA8003-0205PK 91.16 CAD
CA8003-0205 CA8004-0201 CAAG8001-0201 CAAG8003-0203 CAAG8004-0204 CAAG8010-0401 CAAG8001-0202 CAAG8001-0203 CAAG8002-0201 CAAG8002-0203 CAAG8002-0204 CAAG8003-0202 CAAG8003-0204 CAAG8004-0202 CAAG8004-0203
Liner O-Rings for Third Party GC Instruments, Agilent Technologies
Seals Sealing Rings
Agilent provides a range of high-quality liner O-rings for various third party GC systems.
  • Liners are sealed in the inlet with fluor elastomer or graphite O-rings
  • Graphite O-rings are used when inlet temperatures exceed 350 °C
  • Fluor elastomer O-rings are easier to replace than graphite O-rings, which deteriorate more readily under certain conditions
Liner O-rings are used to form a proper seal around the outside of the inlet liner between the inlet weldment and the turn-top. O-rings are composed of either graphite or fluorocarbon material, Graphite O-More Product Information