High Intensity UltrAA Lamps, Agilent Technologies

Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5610108100 5610108200 5610108300 5610133400
CAAG5610108100EA 1590.57 CAD
CAAG5610108100 CAAG5610108200 CAAG5610108300 CAAG5610133400
High Intensity UltrAA Lamps, Agilent Technologies
Lights Hollow Cathode Lamps
UltrAA Lamps are high intensity light sources for improved performance with Agilent AA systems.

  • Compatible with all Agilent AA systems, although an upgrade kit may need to be installed, depending on the model
  • Emission intensity is increased by 3 to 5 times, for lower noise
  • Sensitivity increased by up to 40% due to the sharper emission profile, allowing determinations at lower levels
  • Element coded for automatic lamp recognition, preventing errors
  • Proven processing steps ensure optimum purity, stable operation, and long lifetime

Agilent UltrAA lamps are high-intensity, boosted discharge hollow cathode lamps that replace conventional lamps used with AA systems. Lamps use standard lamp current but apply an extra boost discharge to increase the emission intensity. The boost current is supplied by a secondary control module (integrated in the instrument, or supplied from an external module). They provide high spectral purity, and the processing, and unique lamp conditioning procedures ensure minimal lamp warm-up and stable operation.

Certifications: Manufactured in an ISO 9001 environment using the purest cathode materials for, optimal lifetime, economical operation, and superb performance.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR