Cell Holders, Bases & Accessories for UV-Vis & UV-Vis-NIR, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
08451-60104 110059900 210125300 7910033500 7910038700 7910047600
CA08451-60104EA 881.85 CAD
CA08451-60104 CAAG110059900 CAAG210125300 CAAG7910033500 CAAG7910038700 CAAG7910047600
Cell Holders, Bases & Accessories for UV-Vis & UV-Vis-NIR, Agilent Technologies
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Standard, cylindrical, long and variable pathlength cell holders and bases for Agilent UV-Vis systems.
  • Standard cell holders replace those supplied with the instrument
  • Standard cell holder ensures quick and precise cell positioning for each measurement
  • Standard cell holders include cell lifter and adjustment screw
  • Long pathlength cell holders have fixed mounting positions that enable rectangular cells up to 100 mm pathlength to be used
  • Variable pathlength cell holders enable rectangular cells with pathlengths from 5 to 50 mm to be used
  • Cylindrical cell holder allows use of cylindrical cells with up to 100 mm pathlength
  • Test tube holder allows use of 16 mm od test tubes in the sample compartment instead of conventional cuvettes
Agilent offers a range of cell holders, providing you with the ability to analyze a wide variety of samples. Standard, cylindrical, long pathlength, and variable pathlength cell holders, bases and test tube holders are all available for Agilent UV-Vis and UV-Vis-NIR systems.