General Purpose Septa for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5080-8728-50 5080-8895 5181-3383-50 5183-4459 9301-1031 5080-8896-50 5181-1260 5080-8894-100 5181-3383-100 5190-3978 5080-8728-100 5181-1261 5181-1283-100 5181-1283-50 8010-0249 8010-0250 8010-0252 8010-0253 8010-0257 8010-0261 8010-0262 8010-0263
CA5080-8728-50PK 126.5 CAD
CA5080-8728-50 CA5080-8895 CA5181-3383-50 CA5183-4459 CA9301-1031 CA76514-716 CA76514-732 CA50808894100 CA51813383100 CAAG5190-3978 CAAG5080872810 CAAG5181-1261 CAAG5181128310 CAAG5181128350 CAAG8010-0249 CAAG8010-0250 CAAG8010-0252 CAAG8010-0253 CAAG8010-0257 CAAG8010-0261 CAAG8010-0262 CAAG8010-0263
General Purpose Septa for GC, Agilent Technologies
Septa
Agilent general-purpose septa are made to withstand over 200 automatic injections at an injection port temperature of 350°C.
  • Low bleed for reduced and increased productivity
  • Ability to withstand more than 200 injections at elevated injection port temperatures
  • Easy penetration yet resistant to coring and leaking from multiple injections
  • Blister packaging for cleanliness and convenience
Agilent’s general-purpose septa are made from an enhanced injection-molded silicone rubber material. The septa material, dark red or gray in color, is specified toMore Product Information