CrossLab Syringes for Waters HPLC Autosamplers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
8005-0419 8010-0445 8005-0420 8010-0442 8010-0443 8010-0444 8010-0467
CAAG8005-0419EA 391.64 CAD
CAAG8005-0419 CAAG8010-0445 CAAG8005-0420 CAAG8010-0442 CAAG8010-0443 CAAG8010-0444 CAAG8010-0467
CrossLab Syringes for Waters HPLC Autosamplers, Agilent Technologies
Syringes Syringes for Chromatography
Agilent syringes for Waters autosamplers ensure accurate and effective sampling.
  • Accuracy to within ±1% of nominal volume with a precision of 1% at 80% of the total volume
  • A chemically inert fluid path of stainless steel, borosilicate Type I glass, or PTFE
  • Robust design for long lifetime;PTFE Luer lock, removable, and chem syringe terminations are available
The Agilent HPLC syringe portfolio for Waters autosamplers contains a broad selection of syringe styles and volumes to provide what you need for accurate and effective sampling. From autosamplerMore Product Information