CrossLab Manual Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5190-1463 5190-1483 5190-1490 5190-1497 5190-1510 5190-1517 5190-1523 5190-1528 G4277-80120 5190-0464 5190-1464 5190-1465 5190-1471 5190-1479 5190-1487 5190-1488 5190-1489 5190-1491 5190-1493 5190-1498 5190-1500 5190-1503 5190-1507 5190-1529 5190-1530 5190-1531 5190-1532 5190-1457 5190-1460 5190-1466 5190-1472 5190-1475 5190-1476 5190-1481 5190-1495 5190-1502 5190-1504 5190-1506 5190-1509 5190-1511 5190-1513 5190-1514 5190-1516 5190-1518 5190-1519 5190-1521 5190-1524 5190-1525 5190-1527 5190-1533 5190-1534 5190-1535 5190-1536 5190-1537 5190-1539 5190-1540 5190-1541 5190-1542 5190-1543 5190-1544 5190-1545 5190-1547
CA5190-1463EA 154.43 CAD
CA5190-1463 CA5190-1483 CA5190-1490 CA5190-1497 CA5190-1510 CA5190-1517 CA5190-1523 CA5190-1528 CAG4277-80120 CAAG5190-0464 CAAG5190-1464 CAAG5190-1465 CAAG5190-1471 CAAG5190-1479 CAAG5190-1487 CAAG5190-1488 CAAG5190-1489 CAAG5190-1491 CAAG5190-1493 CAAG5190-1498 CAAG5190-1500 CAAG5190-1503 CAAG5190-1507 CAAG5190-1529 CAAG5190-1530 CAAG5190-1531 CAAG5190-1532 CAAG5190-1457 CAAG5190-1460 CAAG5190-1466 CAAG5190-1472 CAAG5190-1475 CAAG5190-1476 CAAG5190-1481 CAAG5190-1495 CAAG5190-1502 CAAG5190-1504 CAAG5190-1506 CAAG5190-1509 CAAG5190-1511 CAAG5190-1513 CAAG5190-1514 CAAG5190-1516 CAAG5190-1518 CAAG5190-1519 CAAG5190-1521 CAAG5190-1524 CAAG5190-1525 CAAG5190-1527 CAAG5190-1533 CAAG5190-1534 CAAG5190-1535 CAAG5190-1536 CAAG5190-1537 CAAG5190-1539 CAAG5190-1540 CAAG5190-1541 CAAG5190-1542 CAAG5190-1543 CAAG5190-1544 CAAG5190-1545 CAAG5190-1547
CrossLab Manual Syringes, Agilent Technologies
Syringes Syringes for Chromatography
Agilent syringes for manual injection can be used with any type or brand of manual injection valve.
  • Blunt-tip point style prevents damage to valve's internal parts
  • Can be used with any type or brand of manual injection valve
Agilent syringes for manual injection have a blunt-tip needle to prevent damaging the valve's internal parts. These syringes can be used with any type or brand of manual injection valve. For more help choosing the right syringe for your application, check out our syringe selection tool.
Order Now
SPECIFICATIONS
Environmentally Preferable