CrossLab CTC Autosampler Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G7378-80101 8010-0277 8010-0449 G7378-80100 G7378-80102
CAAGG737880101EA 707.16 CAD
CAAGG737880101 CAAG8010-0277 CAAG8010-0449 CAAGG737880100 CAAGG737880102
CrossLab CTC Autosampler Syringes, Agilent Technologies
Syringes Syringes for Chromatography
Agilent has what you need for precise, effective sample delivery from your CTC GC autosampler.
  • CTC syringes provide precise and effective sample delivery from your CTC autosampler to Agilent GC
  • Handles both liquid and headspace injections.;Compatible with both CombiPAL and PAL3
For precise, effective, sample delivery from your CTC autosampler, Agilent has the right syringe for you. Whether you're performing liquid or headspace injections, or using a CombiPAL or PAL3 autosampler, you'll find what you need among our wide range of syringes for CTCMore Product Information