CrossLab Autosampler Plungers, Agilent Technologies
Supplier: Agilent
CrossLab
5181-3365 5190-1560 4710003100 5190-1572 5181-3365 5190-1559 5190-1564 5190-1574 5190-1562 8010-0359 5190-1573 8010-0359 5190-1565 5190-1559 5190-1561 5190-1574 5190-1565 4710003100 5190-1572 5190-1564 5190-1560 5190-1573 5190-1561 5190-1558 5190-1558 5190-1562
CAAG5190-1574EA 77.74 CAD
CAAG5190-1574 CAAG5190-1573 AG5190-1572 CAAG5190-1572 AG5190-1574 CA8010-0359 AG5190-1573 CAAG5190-1559 AG5190-1558 CAAG5190-1558 AG5190-1559 AG8010-0359 CAAG5190-1562 CAAG5190-1561 AG5190-1561 CAAG5190-1560 AG5190-1560 CAAG4710003100 AG5190-1562 AG5190-1565 AG5190-1564 CA5181-3365 CAAG5190-1565 CAAG5190-1564 AG5181-3365 AG4710003100
CrossLab Autosampler Plungers, Agilent Technologies
Syringe Barrels, Plungers and Pistons
Not ready for a new syringe? Try out our replacement needles and plungers.
  • Available for a variety of syringe makes in a choice of PTFE-tipped or metal fitted plunger to meet your application needs
  • Environmentally friendly packaging, with an improved design that reduces waste
  • Fresh needles help to reduce septum coring and keeps your system working at its full potential
  • Help to maintain a chemically inert fluid path of stainless steel, borosilicate Type I glass, or PTFE
Agilent replacement needles and plungers ensure your syringes continue to function at their highest level of performance. Available in various volumes and styles, you'll be sure to find the product that fits your needs, regardless of syringe make and model. Agilent needles and plungers are manufactured with strict tolerances and intense quality standards. This ensures the product performs the way you expect it to, every single time.