Autosampler Needles and Plungers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1313-87203 G1329-80001 G3160-65306 G1313-87201 G1313-87202 G7129-87200 G7129-87201 G7129-87202 5190-1496 5190-1548 5190-1549 G4277-80125 RF0050T 5181-7442 5181-8811 5182-0830 5190-1461 5190-1473 5190-1477 5190-1478 5190-1482 5190-1538 5190-1550 5190-1571 5190-1581 5190-1582 8001-0005 8001-0007 8001-0014 8002-0005 8010-0367 G1811-23200 RF0050T-S RF0052T
CAG1313-87203EA 307.81 CAD
CAG1313-87203 CAG1329-80001 CAG3160-65306 CA76508-240 CA76508-242 CAG7129-87200 CAG7129-87201 CAG7129-87202 CAAG5190-1496 CAAG5190-1548 CAAG5190-1549 CAAGG427780125 CAAGRF0050T CAAG5181-7442 CAAG5181-8811 CAAG5182-0830 CAAG5190-1461 CAAG5190-1473 CAAG5190-1477 CAAG5190-1478 CAAG5190-1482 CAAG5190-1538 CAAG5190-1550 CAAG5190-1571 CAAG5190-1581 CAAG5190-1582 CAAG8001-0005 CAAG8001-0007 CAAG8001-0014 CAAG8002-0005 CAAG8010-0367 CAAGG181123200 CAAGRF0050T-S CAAGRF0052T CAAGRF0052T-S
Autosampler Needles and Plungers, Agilent Technologies
Syringe Needles
Not ready for a new syringe? Try out our replacement needles and plungers.
  • Available for a variety of syringe makes in a choice of PTFE-tipped or metal fitted plunger to meet your application needs
  • Environmentally friendly packaging, with an improved design that reduces waste
  • Fresh needles help to reduce septum coring and keeps your system working at its full potential
  • Help to maintain a chemically inert fluid path of stainless steel, borosilicate Type I glass, or PTFE
Agilent replacement needles and plungers ensure your syringes continue to function at their highest level of performance. Available in various volumes and styles, you'll be sure to find the product that fits your needs, regardless of syringe make and model. Agilent needles and plungers are manufactured with strict tolerances and intense quality standards. This ensures the product performs the way you expect it to, every single time.