Motic Moticam A Class, Microscope Cameras, National Optical
Supplier: National Optical
D-MOTICAM A1 D-MOTICAM A2 D-MOTICAM A5
470336-478EA 401.3 CAD
470336-478 470336-480 470336-482
Motic Moticam A Class, Microscope Cameras, National Optical
Cameras
Ideal for in-person or distance learning and instruction.
  • sCMOS sensor
  • Fast frame rates
  • MotiConnect software
The Moticam A-Line follows the Motic All-In-One Box methodology. This means that with the included focusable lenses, eyepiece adapters and glass calibration slide, you can convert almost any conventional microscope into a digital microscope.
This line of cameras feature Motic's state-of-the-art cMOS imaging units packed in high quality metal housing. The on-board UVC technology means that you no longer have to search for or install aMore Product Information