CrossLab Pistons and Seals for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1361-22402 0905-1420 0905-1719 5022-2188 5067-5678 5067-5975 5067-4299 5067-6587 G4220-60016 5067-6588 G4204-40005 G4220-60015 G5611-60016 G7131-20009 G7161-60005 G7161-60028
CAG1361-22402EA 721.02 CAD
CAG1361-22402 CA76508-020 CA0905-1719 CA5022-2188 CA5067-5678 CA5067-5975 CAAG5067-4299 CAAG5067-6587 CAAGG422060016 CAAG5067-6588 CAAGG420440005 CAAGG422060015 CAAGG561160016 CAAGG713120009 CAAGG716160005 CAAGG716160028
CrossLab Pistons and Seals for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Pistons and seals designed to ensure your pump delivers the best performance.
  • Sapphire pistons are meticulously cut at just the right angle, making them durable and long lasting
  • Pistons are precisely aligned in their stainless steel holder to minimize wear on the piston and seal
  • Piston seals are spring loaded and engineered to deliver optimal performance over highly dynamic flow and pressure ranges
  • Seals are manufactured from a proprietary polymer blend and feature a spring made from the same high-quality stainless steel that is used in our pump's flow path
Agilent pistons are made from a high-purity, ceramic or monocrystalline sapphire for maximum durability. Agilent seals are designed to fit perfectly around pistons, and are capable of adapting to a wide range of flow rates and pressures. The combination of our piston and seal has undergone extensive testing under temperature stress, with all common HPLC solvents, and in many instruments, and have been shown to yield consistent, reproducible results.