CrossLab Vacuum Pump Supplies for GC/MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
1535-4970 3162-1056 G1099-80039 G3170-80006 392028900 393064601
CA1535-4970EA 258.64 CAD
CA1535-4970 CA3162-1056 CAG1099-80039 CAAGG317080006 CAAG392028900 CAAG393064601
CrossLab Vacuum Pump Supplies for GC/MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A range of vacuum pump supplies to ensure the accuracy and performance of your GC/MS system.
  • Oil mist and exhaust filters for vacuum and rotary vane pumps;Oil for turbo, diffusion, and foreline pumps
  • Further accessories including oil return lines, oil pans, vacuum gauge tubes
Agilent provides a range of vacuum pumps and supplies to ensure the highest levels of accuracy and performance of our GC/MS systems. These supplies include foreline and diffusion pump supplies such as pump oil, oil mist filter kits, and gauge tubes for measuring vacuum. TubingMore Product Information