Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3286-80117 G3286-80118 G8010-60303 G8010-60306 G8010-60308 G8010-60304 G8010-60307 G8010-60335 G8010-60348 9910057100 9910097500 9910105400 G8010-60333 G8010-60334 G8410-68261 G8479-68100
CAG3286-80117EA 257.82 CAD
CAG3286-80117 CAG3286-80118 CAG8010-60303 CAG8010-60306 CAG8010-60308 CAAGG801060304 CAAGG801060307 CAAGG801060335 CAAGG801060348 CAAG9910057100 CAAG9910097500 CAAG9910105400 CAAG9910123600 CAAG9910123800 CAAG9910123900 CAAG9910124200 CAAG9910124300 CAAG9910124400 CAAG9910124500 CAAGG801060333 CAAGG801060334 CAAGG841068261 CAAGG847968100 CAAGG848560515 CAAGG848560523
Tubing Kits for ICP-OES
Chromatography Supplies Chromatography Spare Parts
Complete tubing kits to match application compatibility for ICP-OES.
  • Convienent tubing kits provide all the compatible tubing needed for your ICP-OES that is compatible with your sample matrix
  • Reduce costs by purchasing a tubing kit vs buying each item individually
Agilent provides convenient tubing kits for handling aqueous samples, semi-volatile organic solvents, volatile organic solvents, ketone-based organic solvents, and aromatic organic solvents for the Agilent ICP-OES instrument. Each kit includes peristaltic pump tubing, drain tubing, waste line, EzyFit connectors, nebulizer capillary tubing, connectors and connecting tubing.