CrossLab Tools and Other Spray Chamber Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
7210027700 7210001600 7210027600
CAAG7210027700EA 53.97 CAD
CAAG7210027700 CAAG7210001600 CAAG7210027600
CrossLab Tools and Other Spray Chamber Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Spray chamber tools help with routine maintenance of the Mark 7 flame atomization system.
  • These tools are required for maintenance of the nebulizer, that is integrated into the nebulizer block of the Mark 7 spray chamber assembly
  • Maintenance tools enable disassembly and removal of the nebulizer components
  • These tools are also required for reassembly of the nebulizer after cleaning or replacement of worn components
Spray chamber tools enable the user to complete routine maintenance on the Mark 7 spray chamber. Maintenance is easy. Special spray chamber tools are required, to remove the nebulizer components which are integrated into the nebulizer block.
The nebulizer capillary extraction tool is required to remove the bush, retaining the nebulizer components, from the nebulizer block.
The venturi extraction tool is required to gently push the venturi through the nebulizer block, and to dislodge the capillary guide.
These tools are also required for reassembly of the nebulizer components into the nebulizer block after cleaning or replacement of worn components.