CrossLab Pump and Maintenance Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5067-6137 G1311-68711 G1312-68711 G4220-20001
CAAG5067-6137KT 19.24 CAD
CAAG5067-6137 CAAGG131168711 CAAGG131268711 CAAGG422020001
CrossLab Pump and Maintenance Kits for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A comprehensive range of kits for maintaining and upgrading HPLC pumps.
  • A lack of preventive maintenance can cause noisy baselines, shorter column life, lower analytical sensitivity, and variable retention times
  • HPLC pumps work at their best with the Agilent preventive maintenance kits
  • Routine pump maintenance should be done on a regular basis to keep your Agilent LC system performing at its optimum
  • By following a regular maintenance routine you can count on maximum uptime and a steady, accurate solvent flow for the life of the pump
Agilent provides a comprehensive range of kits for HPLC pumps. Maintenance kits include the right supplies to be exchange on a regular basis. Accessory kits contain parts to always have on-hand. Upgrade kits allow users to change the original setup.
All parts are manufactured to Agilent's highest levels of quality and have been tested in Agilent instruments to ensure they work perfectly with the rest of the system.