CrossLab Other Tubing for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
2410020500 7910051590 0890-2603 3710024600 3710033400 7910051290 G8410-20012 G8410-20013 G8410-80122 G8410-80123 3710031500
CA2410020500EA 13.46 CAD
CA2410020500 CA7910051590 CAAG0890-2603 CAAG3710024600 CAAG3710033400 CAAG7910051290 CAAGG841020012 CAAGG841020013 CAAGG841080122 CAAGG841080123 CA3710031500
CrossLab Other Tubing for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Non peri-pump tubing for ICP-OES such as drain tubing, and capillary tubing.
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
Agilent has a wide variety of high-quality tubing for your ICP-OES. From ICP-OES drain/waste tubing to nebulizer capillary tubing, Agilent has the tubing to fit your ICP-OES in a material compatible with your matrix.
The reagent uptake capillary tube supply line is fitted with a heavy PTFE sinker to ensure the capillary draws from the base of the container. This convienent capillary line with sinker can be used for rinse and internal standard uptake to minimize air bubbles and refilling requirements.