CrossLab Nebulizers for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1820-65138 G3266-65003 G3266-80004 G3139-65102 G3266-80005 G3139-65100 G3161-80001 G3139-65106 G3139-65108 G3161-80000 G3285-80000 G3285-80001 G3285-80002 G3285-80003
CAG1820-65138EA 2559.39 CAD
CAG1820-65138 CAG3266-65003 CAG3266-80004 CAG3139-65102 CAG3266-80005 CAAGG313965100 CAAGG316180001 CAAGG313965106 CAAGG313965108 CAAGG316180000 CAAGG328580000 CAAGG328580001 CAAGG328580002 CAAGG328580003
CrossLab Nebulizers for ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent has MicroMist and inert PFA concentric nebulizers to suit your ICP-MS application needs.
  • Concentric nebulizers are routinely recommended for use with ICP-MS for all but the highest matrix samples which may cause the inner capillary to become clogged by particles or high concentrations of salt
  • Two types of concentric nebulizer are available: Quartz (recommended for trace analysis of clean samples) and PFA (recommended for semi-conductor process chemicals - providing inertness to HF and self-aspiration)
  • High TDS/matrix samples are tolerated by parallel path nebulizers such as the MiraMist available in PEEK or PTFE for inertness to high acid concentration and solvents
  • A full range of connection accessories including U-series and Ezy Fit sample connectors, tubing, O-rings and other fittings are available for all models, plus nebulizer cleaning tools, manufactured to Agilent's high quality standards
  • Ensure the longevity of your nebulizer through regular cleaning with a 5% HNO₃ (v/v) bath (do not use an ultrasonic bath)
  • Replace the nebulizer when the capillary is broken or the nebulizer is damaged
Agilent has a range of nebulizers and supplies to best suit your ICP-MS application needs. From standard glass or inert PFA concentric nebulizers, robust nebulizers for high TDS samples, to low-flow and self-aspirating nebulizers for your ICP-MS.
MicroMist glass concentric nebulizers and a full range of connection accessories including U-series and Ezy Fit sample connectors, tubing, nebulizer cleaning tools, O-rings and other fittings are available for all models.