CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G8020-60806 G8020-60807 2010095400 2010095900 2010096000 2010101700 2410067590 G8020-60805 G8020-60808
CAG8020-60806EA 429.43 CAD
CAG8020-60806 CAAGG802060807 CAAG2010077500 CAAG2010095400 CAAG2010095600 CAAG2010095900 CAAG2010096000 CAAG2010101700 CAAG2410067590 CAAGG802060805 CAAGG802060808 CAAGG802060809
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
ICP-OES quartz and ceramic injectors for demountable torches.
  • A variety of injector sizes and materials to handle difficult and diverse application demands
  • Removable injectors provide quick change-over without the need for diagrams or multiple o-rings and seals
  • No optimization required with the Agilent fully demountable torch providing consistent results before and after injector replacement regardless of number of analysts
  • Greater productivity Tool free installation simplifies injector maintenence
Agilent ICP-OES injectors for fully demountableMore Product Information