Inert ICP-OES nebulizers including the OneNeb Series 2 nebulizer offering superior performance.
- OneNeb 2 is a direct replacement for standard glass concentric nebulizers and other inert nebulizers - that have a 6 mm od tip
- OneNeb 2 is suitable for use with Agilent ICP-OES and MP-AES systems, and other ICP-OES systems where a 6mm od nebulizer is used
- OneNeb 2 and Miramist are suitable for conventional glass cyclonic and inert spray chambers - that are compatible with nebulizers with a 6 mm od tip
- OneNeb 2 provides improved sensitivity and precision - due to the fine aerosol generated by flow blurring nebulization
- OneNeb 2 offers high efficiency - at both normal and micro sample uptake rates
- Greater tolerance to samples with high Total Dissolved Solids (TDS) content
- Inert construction - ensures the OneNeb that can handle most ICP-OES sample matrices
Agilent ICP-OES inert nebulizers analyze samples with hydrofluoric (HF) acid. HF-resistant nebulizers handle HF digests, unlike standard glass or quartz nebulizers. Available ICP-OES inert nebulizers include the One Neb 2, MiraMist, and V-groove nebulizer. The OneNeb Series 2 nebulizer is universal and highly efficient with superior performance with ICP-OES instruments. The OneNeb series 2 nebulizer has improved sensitivity and precision and greater tolerance to dissolved solids. Its universal design handles most ICP-OES sample matrices, including strong acids like HF and common organic acids