CrossLab Heat Exchanger for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1316-80002 G1316-80003 G7116-60015 G1316-80022 G7116-60021 G7116-60041 G7116-60051 G5616-81000 G7116-60009 G7116-60061 G1316-80004 G1316-80012 G7116-60031 G7116-60071 G7116-60081 G7132-68380
CAG1316-80002EA 466.92 CAD
CAG1316-80002 CAG1316-80003 CAG7116-60015 CAG1316-80022 CAG7116-60021 CAG7116-60041 CAG7116-60051 CAAGG561681000 CAAGG711660009 CAAGG711660061 CAAGG131680004 CAAGG131680012 CAAGG711660031 CAAGG711660071 CAAGG711660081 CAAGG713268380
CrossLab Heat Exchanger for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Heat exchangers in different volumes, materials, and designs to fit your column compartment.
  • Different inner diameters (0.12 mm, 0.17 mm) and internal volumes (1.6 µl, 1.0 µl, 6.0 µl) available to meet different separation needs
  • InfinityLab Quick-Connect heat exchangers with improved usability and robustness for hassle-free pre-column heating
  • Perfect fit to the thermostatted column compartment (TCC) or multicolumn compartment (MCT) in your LC system
Agilent provides a variety of heat exchangers in different volumes, materials, and designs, for pre-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable