CrossLab Gas and Liquid Sampling Valve Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0101-0299 0101-0303 0101-0584 07675-80050 1530-2163 18900-20300 5062-9508 5181-7459 18900-21000 18900-20250 18900-20280 18900-20281 18900-20850 18900-80255 5062-9509 5062-9510 5062-9511 5181-7460 G1540-30024 G1540-30025 G1540-30026 G3440-20004 G3440-20033 G3440-20035 G3440-20037 G3440-60033 G3440-60035 G3440-60234 G3440-60236 G3440-60300 G3440-60310 G3440-60336 G3440-60600 G3440-81665 G3480-20002 VLZF1S610 G1540-30027 G1540-30028
CA0101-0299EA 104.41 CAD
CA0101-0299 CA0101-0303 CA0101-0584 CA07675-80050 CA1530-2163 CA18900-20300 CA5062-9508 CA5181-7459 CAAG1890021000 CAAG1890020250 CAAG1890020280 CAAG1890020281 CAAG1890020850 CAAG1890080255 CAAG5062-9509 CAAG5062-9510 CAAG5062-9511 CAAG5181-7460 CAAGG154030024 CAAGG154030025 CAAGG154030026 CAAGG344020004 CAAGG344020033 CAAGG344020035 CAAGG344020037 CAAGG344060033 CAAGG344060035 CAAGG344060234 CAAGG344060236 CAAGG344060300 CAAGG344060310 CAAGG344060336 CAAGG344060600 CAAGG344081665 CAAGG348020002 CAAGVLZF1S610 AGG1540-30027 AGG1540-30028
CrossLab Gas and Liquid Sampling Valve Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent’s portfolio of gas and liquid sampling valve supplies are designed for complex applications.
  • Gas sampling valves are available in Hastelloy C, a robust metal alloy ideal for corrosive or polar analytes, or in Nitrionic 60 stainless steel, which is the standard choice for most general applications
  • Gas sampling valves are available as 4, 6, 10, or 14 port valves
  • Sample loops for gas sampling valves are available from 0.025 μl to 10 ml
  • General purpose liquid sampling valves are stainless steel, and available with 0.2 μl, 0.5 μl, or 1 μl
Order Now
SPECIFICATIONS
Environmentally Preferable