CrossLab FTIR Instrument and Imaging System Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
899-1794 925-0128 883-9750 G8180-67610 PIKE-161-1903 PIKE-162-1320 SPECAC04090
CAAG899-1794EA 757.02 CAD
CAAG899-1794 CAAG925-0128 CAAG883-9750 CAAGG818067610 CAGPIKE1611903 CAGPIKE1621320 CAGSPECAC04090
CrossLab FTIR Instrument and Imaging System Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Optimize performance and maximize efficiency with Cary FTIR instrument and imaging system supplies.
  • Use the coarse gold mirror reference cap for standard diffuse samples or external reflectance
  • Use the smooth gold mirror reference cap for external reflectance (45° or grazing angle)
  • Use the coarse silver mirror reference cap for extremely diffuse samples
  • Use the smooth silver mirror reference cap for highly reflective samples typically requiring grazing angle measurements
An external reflectance measurement using FTIR requires a reference sample toMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable