CrossLab FLD Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
2140-0600 9301-0407 9301-1446 5062-2462 G1321-60005 G1321-60015 G5615-60005
CA2140-0600EA 1594.31 CAD
CA2140-0600 CA9301-0407 CA9301-1446 CA5062-2462 CA76515-216 CAAGG132160015 CAAGG561560005
CrossLab FLD Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A range of high-quality replacement supplies and components for fluorescence detectors (FLD).
  • Agilent quality parts are designed, tested, and manufactured with the same attention to detail you expect from Agilent instruments;Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise.
Agilent provides a range of high-quality replacement supplies and components for fluorescence detectors (FLD). The range of consumables includes Xenon flash detector lamps, flow cells, and a variety of tubing and capillary fittings.More Product Information