CrossLab Degasser Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0100-1710 5022-2155 5063-6598 5063-6599 8710-1930 9301-1337 G1322-67300 G1322-68705 5062-2483 5042-8922 G1379-67310 5067-4798 5067-5378 5067-5380 5067-5383 5067-5387 5067-5388 5067-6624
CA0100-1710EA 20.66 CAD
CA0100-1710 CA5022-2155 CA5063-6598 CA5063-6599 CA8710-1930 CA9301-1337 CAG1322-67300 CAG1322-68705 CA76508-052 CA5042-8922 CAG1379-67310 CA5067-4798 CA5067-5378 CAAG5067-5380 CAAG5067-5383 CAAG5067-5387 CAAG5067-5388 CAAG5067-6624
CrossLab Degasser Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Extensive range of supplies for degasser instruments to remove dissolved oxygen from HPLC solvents.
  • Fast and easy flushing, purging, priming, and solvent exchange;High degassing efficiency for the most demanding applications
  • Highly reproducible analyses
  • Very low baseline noise
Agilent provides an extensive range of supplies for degasser instruments to optimize the HPLC results by removing any trace of dissolved oxygen in HPLC solvents. The Agilent vacuum degasser is recommended for maximum sensitivity in the low-UV wavelength range, for high injection precision, for high retention time reproducibility, and for low flow rate methods.